Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgef10l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001112722.1 | NP_001106193.1 | 9 | 948 | ||
NM_001112723.1 | NP_001106194.1 | 9 | 921 | |||
NM_001290803.1 | NP_001277732.1 | 10 | 1047 | |||
NM_172415.3 | NP_766003.3 | 10 | 1038 | |||
GenBank mRNA | AK028570.1 | - | 3 | 363 | ||
AK028648.1 | - | 9 | 931 | |||
AK129408.1 | - | 9 | 921 | |||
AK203541.1 | - | 3 | 245 | |||
BB842000.1 | - | 2 | 165 | |||
BC034061.1 | - | 3 | 184 | |||
BC059220.1 | - | 10 | 1026 | 6417641 |