Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ostm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172416.3 | NP_766004.1 | 3 | 628 | ||
GenBank mRNA | AF533890.1 | - | 3 | 627 | ||
AK004546.1 | - | 3 | 605 | |||
AK044897.1 | - | 3 | 627 | |||
AK050513.1 | - | 3 | 602 | |||
AK053664.1 | - | 3 | 578 | |||
BC057635.1 | - | 3 | 566 | 4910872 |