Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pif1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172453.3 | NP_766041.1 | 2 | 1301 | ||
GenBank mRNA | AK028460.1 | - | 2 | 1302 | ||
AK049353.1 | - | 2 | 1303 | |||
AK154072.1 | - | 2 | 1314 | |||
AY100322.1 | - | 3 | 1014 | |||
AY498715.1 | - | 2 | 1301 | |||
BC046611.1 | - | 3 | 939 | 6396273 | ||
BC060108.1 | - | 2 | 1292 |