Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172436.3 | NP_766024.1 | 4, 5 | 353 | ||
GenBank mRNA | AK019150.1 | - | 4, 5 | 355 | ||
AK030595.1 | - | 4, 5 | 383 | |||
AK083156.1 | - | 4, 5 | 354 | |||
AK144958.1 | - | 4, 5 | 387 | |||
AK147948.1 | - | 4, 5 | 368 | |||
AK208080.1 | - | 1, 2 | 21 | |||
AK208704.1 | - | 1, 2 | 21 | |||
BC013845.1 | - | 4, 5 | 332 | |||
BC024952.1 | - | 4, 5 | 548 | |||
BC060505.1 | - | 4, 5 | 340 | 6311836 |