Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172448.3 | NP_766036.2 | 10 | 4266 | ||
GenBank mRNA | AK028750.1 | - | 9 | 3625 | ||
BC011216.1 | - | 5 | 1073 | |||
BC029717.1 | - | 2 | 2642 | |||
BC037465.1 | - | 2 | 2706 | |||
CF747567.1 | - | 1 | 441 |