Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pif1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172453.3 | NP_766041.1 | 12 | 3385 | ||
GenBank mRNA | AK049353.1 | - | 12 | 3387 | ||
AK154072.1 | - | 12 | 3431 | |||
AY100322.1 | - | 13 | 3098 | |||
AY498715.1 | - | 12 | 3385 | |||
BC060108.1 | - | 10 | 3596 |