Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Adamts18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172466.2 | NP_766054.2 | 17, 18 | 3045 | ||
GenBank mRNA | AK036137.1 | - | 14, 15 | 2124 | ||
AK053851.1 | - | 16, 17 | 3157 | |||
AK140876.1 | - | 17, 18 | 2821 | |||
AK163023.1 | - | 2, 3 | 273 | |||
BC094674.1 | - | 17, 18 | 2700 | 30548647 | ||
BC144816.1 | - | 15, 16 | 2303 | |||
BC144817.1 | - | 14, 15 | 2175 |