Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Dse | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172508.2 | NP_766096.1 | 2 | 682 | ||
GenBank mRNA | AK031737.1 | - | 2 | 369 | ||
AK045065.1 | - | 2 | 589 | |||
AK150737.1 | - | 2 | 846 | |||
AK153326.1 | - | 2 | 355 | |||
BC053095.1 | - | 2 | 682 | 6401066 |