Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Engase | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172573.2 | NP_766161.1 | 1, 2 | 153 | ||
GenBank mRNA | AK048667.1 | - | 1, 2 | 155 | ||
AK167142.1 | - | 1, 2 | 125 | |||
BC060640.1 | - | 1, 2 | 130 | |||
BC120923.1 | - | 1, 2 | 155 | |||
BC120924.1 | - | 1, 2 | 155 | 40097903 |