Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ybey | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172550.4 | NP_766138.3 | 2 | 533 | ||
GenBank mRNA | AK037727.1 | - | 2 | 260 | ||
AK051909.1 | - | 1 | 670 | |||
AK136543.1 | - | 2 | 284 | |||
BC057308.1 | - | 2 | 533 | 5703033 |