Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Papolg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172555.2 | NP_766143.2 | 2, 3 | 419 | ||
GenBank mRNA | AK035810.1 | - | 2, 3 | 402 | ||
AK042629.1 | - | 2, 3 | 386 | |||
AK139065.1 | - | 2, 3 | 386 | |||
BC049163.1 | - | 2, 3 | 365 | |||
BC059271.1 | - | 2, 3 | 413 | 2811338 | ||
DQ103505.1 | - | 2, 3 | 225 |