Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167976.1 | NP_001161448.1 | 4 | 221 | ||
NM_172577.3 | NP_766165.2 | 3 | 664 | |||
GenBank mRNA | AK036988.1 | - | 3 | 404 | ||
AK041694.1 | - | 3 | 665 | |||
AK044945.1 | - | 3 | 167 | |||
AK045180.1 | - | 3 | 490 | |||
AK167535.1 | - | 3 | 372 | |||
BC057980.1 | - | 3 | 488 | 5042653 | ||
EU180236.1 | - | 4 | 221 |