Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Larp4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172585.2 | NP_766173.1 | 3, 4 | 463 | ||
GenBank mRNA | AK042491.1 | - | 3, 4 | 461 | ||
AK080953.1 | - | 3, 4 | 487 | |||
AK160759.1 | - | 3, 4 | 464 | |||
BC059218.1 | - | 3, 4 | 443 | |||
BC078640.1 | - | 3, 4 | 386 | 6506267 | ||
BC086626.1 | - | 3, 4 | 443 |