Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Astl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291003.1 | NP_001277932.1 | 9 | 2077 | ||
NM_172539.3 | NP_766127.1 | 9 | 2208 | |||
GenBank mRNA | AK033037.1 | - | 9 | 2208 | ||
BC064729.1 | - | 9 | 2069 | 30476764 | ||
DV662582.1 | - | 1 | 481 |