Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rundc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172566.4 | NP_766154.2 | 5 | 1966 | ||
GenBank mRNA | AK086362.1 | - | 5 | 1967 | ||
AK157156.1 | - | 5 | 1960 | |||
BC119592.1 | - | 5 | 1967 | 40059040 | ||
BC119593.1 | - | 5 | 1967 | 40059041 |