Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Clic5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172621.2 | NP_766209.1 | 6 | 2366 | ||
GenBank mRNA | AK030659.1 | - | 1 | 445 | ||
AK046522.1 | - | 6 | 2357 | |||
AK156849.1 | - | 6 | 2214 | |||
AK167584.1 | - | 1 | 387 |