Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ldlrad4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172631.3 | NP_766219.2 | 6 | 2035 | ||
GenBank mRNA | AK052338.1 | - | 6 | 2036 | ||
BC096371.1 | - | 5 | 1796 | 3583946 |