Gene Symbol: Hectd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163471.1 | NP_001156943.1 | 21 | 2464 | ||
NM_172637.3 | NP_766225.2 | 21 | 2461 | |||
GenBank mRNA | AK029549.1 | - | 21 | 2321 | ||
AK035511.1 | - | 7 | 691 | |||
AK142777.1 | - | 19 | 2288 | |||
BC137671.1 | - | 21 | 2435 | 8860690 | ||
BC144815.1 | - | 21 | 2435 | 9053363 |