Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc2a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177627.1 | NP_001171098.1 | 9 | 1786 | ||
NM_172659.2 | NP_766247.2 | 10 | 1948 | |||
GenBank mRNA | AK079650.1 | - | 10 | 1893 | ||
AK089246.1 | - | 10 | 1940 | |||
AK137441.1 | - | 9 | 2163 | |||
AK150104.1 | - | 10 | 1901 | |||
AK166312.1 | - | 10 | 1906 | |||
BC025498.1 | - | 7 | 2531 | |||
BC058210.1 | - | 9 | 2157 |