Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172723.4 | NP_766311.2 | 4 | 454 | ||
GenBank mRNA | AK076754.1 | - | 3 | 440 | ||
AK076768.1 | - | 4 | 211 | |||
BC056934.1 | - | 3 | 180 | |||
BC079634.1 | - | 3 | 248 | |||
BC144874.1 | - | 3 | 199 | |||
CX242691.1 | - | 4 | 454 |