Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap5z1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172725.2 | NP_766313.2 | 10 | 1396 | ||
GenBank mRNA | AK040924.1 | - | 8 | 948 | ||
AK082759.1 | - | 2 | 223 | |||
AK129135.1 | - | 7 | 2594 | |||
AK146431.1 | - | 10 | 1274 | |||
AK152402.1 | - | 10 | 1274 | |||
AK153787.1 | - | 9 | 2915 | |||
BC057548.1 | - | 10 | 1251 | |||
BC144755.1 | - | 8 | 927 |