Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172739.4 | NP_766327.3 | 7 | 5754 | ||
GenBank mRNA | AK032559.1 | - | 1 | 507 | ||
AK136766.1 | - | 6 | 2428 | |||
AK147326.1 | - | 7 | 5754 | |||
AK147688.1 | - | 7 | 5754 | |||
AK173242.1 | - | 6 | 4556 | |||
BC035541.1 | - | 6 | 1817 | |||
BC057306.1 | - | 1 | 766 | |||
BC066142.1 | - | 1 | 880 | |||
BC111038.1 | - | 1 | 762 | |||
BC139459.1 | - | 8 | 5532 | 8862481 | ||
BC139461.1 | - | 8 | 5391 | 8862483 |