Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 9330182L06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172706.3 | NP_766294.2 | 3 | 437 | ||
GenBank mRNA | AK034358.1 | - | 3 | 469 | ||
AK036245.1 | - | 3 | 513 | |||
AK043026.1 | - | 3 | 501 | |||
AK045969.1 | - | 3 | 471 | |||
AK048738.1 | - | 3 | 457 | |||
AK079644.1 | - | 2 | 368 | |||
AK081041.1 | - | 3 | 482 | |||
AK082474.1 | - | 2 | 382 | |||
AK133610.1 | - | 3 | 438 | |||
AK134808.1 | - | 3 | 498 | |||
AK161902.1 | - | 3 | 498 | |||
AK165601.1 | - | 3 | 491 | |||
BC132290.1 | - | 3 | 413 | 40130567 | ||
BC138012.1 | - | 3 | 413 | 8861031 |