Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sel1l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172710.3 | NP_766298.2 | 5 | 1184 | ||
GenBank mRNA | AK030487.1 | - | 5 | 1049 | ||
AK037220.1 | - | 5 | 1048 | |||
AK122362.1 | - | 5 | 1184 | |||
BC094607.1 | - | 4 | 729 | 6837048 | ||
BC117915.1 | - | 4 | 685 | 40097358 | ||
BC117916.1 | - | 4 | 687 | 40097359 |