Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CASP8 SIRNA Assays ›
Gene Symbol:
CASP8
Gene Name:
caspase 8
Gene Aliases:
ALPS2B, CAP4, Casp-8, FLICE, MACH, MCH5
Chromosome Location:
Chr.2: 201233443 - 201287711 on Build GRCh38
UniGene ID:
Hs.599762
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
9478
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CASP8
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001080124.1
NP_001073593.1
3
341
NM_001080125.1
NP_001073594.1
2
484
NM_001228.4
NP_001219.2
3
409
NM_033355.3
NP_203519.1
3
315
NM_033356.3
NP_203520.1
2
246
NM_033358.3
NP_203522.1
3
409
NR_111983.1
2
484
XM_005246885.1
XP_005246942.1
2
304
XM_005246886.1
XP_005246943.1
2
230
XM_005246887.1
XP_005246944.1
3
382
XM_005246888.1
XP_005246945.1
3
324
XM_005246889.1
XP_005246946.1
2
201
XM_005246890.3
XP_005246947.1
2
208
XM_005246891.4
XP_005246948.1
2
448
XM_005246892.1
XP_005246949.1
2
201
XM_005246893.2
XP_005246950.1
2
304
XM_005246894.3
XP_005246951.1
3
379
XM_005246895.2
XP_005246952.1
2
304
XM_006712789.1
XP_006712852.1
3
341
XM_006712790.3
XP_006712853.1
2
220
XM_006712793.2
XP_006712856.1
3
382
GenBank mRNA
AA312258.1
-
2
227
AB451282.1
-
1
106
AB451414.1
-
1
106
AF009620.1
-
1
106
AF380342.1
-
2
191
AF422926.1
-
2
241
AF422927.1
-
3
329
AF422928.1
-
3
303
AF422929.1
-
2
371
AK310630.1
-
3
382
AL601594.1
-
2
247
BC028223.2
-
1
133
BC068050.1
-
2
196
6421760
BX395849.2
-
2
195
CB990598.1
-
3
326
DA420056.1
-
3
343
EU168332.1
-
1
106
EU670044.1
-
1
106
JN657192.1
-
1
68
U58143.1
-
1
106
U60520.1
-
3
362
X98172.1
-
2
397
X98173.1
-
2
178
X98176.1
-
3
295
X98177.1
-
1
106
X98178.1
-
1
106
Back To Top
More Information
Panther Classification:
Molecular Function -
protease
Gene Ontology Categories:
Function(s)
Process(es)
proteolysis
apoptotic process
activation of cysteine-type endopeptidase activity involved in apoptotic process
cellular component disassembly involved in execution phase of apoptosis
cell surface receptor signaling pathway
response to cold
regulation of tumor necrosis factor-mediated signaling pathway
regulation of necrotic cell death
natural killer cell activation
macrophage differentiation
response to cobalt ion
response to estradiol
response to lipopolysaccharide
response to tumor necrosis factor
TRIF-dependent toll-like receptor signaling pathway
TRAIL-activated apoptotic signaling pathway
suppression by virus of host cysteine-type endopeptidase activity involved in apoptotic process
T cell activation
B cell activation
positive regulation of I-kappaB kinase/NF-kappaB signaling
negative regulation of I-kappaB kinase/NF-kappaB signaling
response to ethanol
positive regulation of macrophage differentiation
positive regulation of proteolysis
response to antibiotic
protein heterooligomerization
proteolysis involved in cellular protein catabolic process
syncytiotrophoblast cell differentiation involved in labyrinthine layer development
nucleotide-binding oligomerization domain containing signaling pathway
cellular response to mechanical stimulus
cellular response to organic cyclic compound
death-inducing signaling complex assembly
apoptotic signaling pathway
extrinsic apoptotic signaling pathway
execution phase of apoptosis
activation of cysteine-type endopeptidase activity
activation of cysteine-type endopeptidase activity involved in apoptotic signaling pathway
positive regulation of protein insertion into mitochondrial membrane involved in apoptotic signaling pathway
regulation of extrinsic apoptotic signaling pathway via death domain receptors
negative regulation of extrinsic apoptotic signaling pathway via death domain receptors
cysteine-type endopeptidase activity
death receptor binding
tumor necrosis factor receptor binding
protein binding
peptidase activity
cysteine-type peptidase activity
ubiquitin protein ligase binding
protein complex binding
death effector domain binding
identical protein binding
scaffold protein binding
cysteine-type endopeptidase activity involved in apoptotic process
cysteine-type endopeptidase activity involved in apoptotic signaling pathway
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-xql29:80/10.239.165.153:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0