Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Zc3h12d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172785.3 | NP_766373.2 | 3 | 570 | ||
GenBank mRNA | AK052806.1 | - | 3 | 572 | ||
BB661745.1 | - | 3 | 571 | |||
BC112398.1 | - | 2 | 352 | 40062049 |