Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zc3hc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172735.2 | NP_766323.1 | 11 | 1517 | ||
GenBank mRNA | AJ537495.1 | - | 10 | 1465 | ||
AK004251.1 | - | 1 | 501 | |||
AK088527.1 | - | 11 | 1518 | |||
AK146228.1 | - | 10 | 1488 | |||
AK151694.1 | - | 10 | 1469 | |||
AK167465.1 | - | 10 | 1474 | |||
AK167992.1 | - | 10 | 1488 | |||
AK168071.1 | - | 10 | 1483 | |||
BC024560.1 | - | 4 | 489 | |||
BC037445.1 | - | 10 | 1427 | |||
BC050141.1 | - | 10 | 1466 | 30011212 |