Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankle1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172756.2 | NP_766344.2 | 9 | 1724 | ||
GenBank mRNA | AK049395.1 | - | 7 | 2031 | ||
AK080768.1 | - | 7 | 2037 | |||
AK083552.1 | - | 7 | 2025 | |||
AK087444.1 | - | 7 | 2043 | |||
BC094316.1 | - | 9 | 1681 | |||
BC145659.1 | - | 5 | 949 |