Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mthfsd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172761.3 | NP_766349.1 | 8 | 1911 | ||
GenBank mRNA | AK083317.1 | - | 8 | 1852 | ||
BC023782.1 | - | 8 | 1888 | |||
BC026652.1 | - | 7 | 1787 | |||
BC052066.1 | - | 7 | 3194 | 5708660 |