Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDH13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001220488.1 | NP_001207417.1 | 3 | 551 | ||
NM_001220489.1 | NP_001207418.1 | 2 | 445 | |||
NM_001220490.1 | NP_001207419.1 | 2 | 445 | |||
NM_001220491.1 | NP_001207420.1 | 2 | 445 | |||
NM_001220492.1 | NP_001207421.1 | 2 | 445 | |||
NM_001257.4 | NP_001248.1 | 2 | 445 | |||
XM_017022848.1 | XP_016878337.1 | 3 | 468 | |||
XM_017022849.1 | XP_016878338.1 | 3 | 513 | |||
GenBank mRNA | AK295886.1 | - | 2 | 275 | ||
AK298612.1 | - | 2 | 271 | |||
AK304733.1 | - | 3 | 306 | |||
AK307862.1 | - | 2 | 272 | |||
AU123178.1 | - | 2 | 270 | |||
BC012615.1 | - | 2 | 203 | |||
BC028624.2 | - | 2 | 237 | 4825734 | ||
BC030653.2 | - | 2 | 237 | 4811561 | ||
BI756586.1 | - | 2 | 248 | |||
CV024366.1 | - | 2 | 146 | |||
DC376661.1 | - | 2 | 445 | |||
EU190357.1 | - | 2 | 272 | |||
EU190358.1 | - | 2 | 272 | |||
L34058.1 | - | 2 | 590 | |||
U59288.1 | - | 2 | 362 |