Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp462 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172867.3 | NP_766455.2 | 10 | 7181 | ||
GenBank mRNA | AK031378.1 | - | 5 | 692 | ||
AK173266.1 | - | 8 | 1494 | |||
BC099692.1 | - | 5 | 778 | |||
DQ355518.1 | - | 9 | 6878 |