Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp462 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172867.3 | NP_766455.2 | 13 | 8470 | ||
GenBank mRNA | AK031378.1 | - | 8 | 1981 | ||
AK035041.1 | - | 1 | 476 | |||
AK173266.1 | - | 11 | 2784 | |||
DQ355518.1 | - | 12 | 8167 |