Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps13a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173028.4 | NP_766616.2 | 48, 49 | 6925 | ||
GenBank mRNA | AB115421.1 | - | 48, 49 | 6920 | ||
AK029649.1 | - | 2, 3 | 680 | |||
AK030073.1 | - | 1, 2 | 149 | |||
AK135983.1 | - | 5, 6 | 1079 | |||
AK220362.1 | - | 1, 2 | 342 | |||
BC062130.1 | - | 4, 5 | 823 | |||
BC096768.1 | - | 2, 3 | 519 | |||
BC145012.1 | - | 48, 49 | 6814 |