Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspan12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173007.3 | NP_766595.1 | 4, 5 | 493 | ||
GenBank mRNA | AK033554.1 | - | 3, 4 | 328 | ||
AK050737.1 | - | 4, 5 | 494 | |||
BC065152.1 | - | 3, 4 | 304 | 6808971 | ||
BC068240.1 | - | 3, 4 | 355 | 3674585 |