Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APBB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164.4 | NP_001155.1 | 2 | 653 | ||
NM_145689.2 | NP_663722.1 | 2 | 653 | |||
NR_047512.2 | 2 | 653 | ||||
XM_006718209.3 | XP_006718272.2 | 2 | 688 | |||
XM_006718210.3 | XP_006718273.2 | 2 | 738 | |||
XM_006718211.1 | XP_006718274.1 | 2 | 894 | |||
XM_006718212.3 | XP_006718275.2 | 2 | 688 | |||
XM_006718213.1 | XP_006718276.1 | 2 | 894 | |||
XM_006718214.3 | XP_006718277.2 | 2 | 688 | |||
XM_006718215.3 | XP_006718278.3 | 2 | 1011 | |||
XM_017017639.1 | XP_016873128.1 | 2 | 894 | |||
XM_017017640.1 | XP_016873129.1 | 2 | 894 | |||
XM_017017641.1 | XP_016873130.1 | 2 | 894 | |||
GenBank mRNA | AF394214.1 | - | 2 | 612 | ||
BC010854.2 | - | 2 | 607 | 3905603 | ||
L77864.1 | - | 2 | 612 |