Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xkr6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173393.2 | NP_775569.2 | 3 | 1036 | ||
GenBank mRNA | AY534252.1 | - | 3 | 742 | ||
BC024502.1 | - | 3 | 592 | 4983341 |