Gene Symbol: Cypt7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039943.2 | NP_001035032.1 | 2 | 470 | ||
GenBank mRNA | AM040456.1 | - | 2 | 470 |
Gene Symbol: Cypt10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039944.2 | NP_001035033.1 | 1 | 428 | ||
GenBank mRNA | AM040459.1 | - | 1 | 428 |
Gene Symbol: Cypt4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173412.2 | NP_775588.2 | 1 | 441 | ||
GenBank mRNA | AF463501.1 | - | 1 | 441 | ||
AK005791.1 | - | 1 | 434 | |||
AM040453.1 | - | 1 | 425 | |||
BC048530.1 | - | 1 | 430 | |||
BC060952.1 | - | 1 | 430 | 6772242 | ||
BC147783.1 | - | 1 | 525 | 9087502 |