Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fkrp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173430.2 | NP_775606.1 | 3 | 1456 | ||
GenBank mRNA | AJ511806.1 | - | 3 | 1439 | ||
AK080624.1 | - | 1 | 1266 | |||
AK135089.1 | - | 3 | 1456 | |||
AK156156.1 | - | 3 | 1455 | |||
AK166068.1 | - | 3 | 1458 | |||
AK168308.1 | - | 3 | 1454 | |||
AK189687.1 | - | 1 | 53 | |||
BC053072.1 | - | 3 | 1439 | 5686286 |