Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ephb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168296.1 | NP_001161768.1 | 3 | 683 | ||
NM_173447.3 | NP_775623.3 | 3 | 683 | |||
GenBank mRNA | AK033966.1 | - | 3 | 683 | ||
AK036148.1 | - | 3 | 371 | |||
AK036211.1 | - | 3 | 682 | |||
AK039530.1 | - | 3 | 684 | |||
AK135018.1 | - | 3 | 649 | |||
BC057301.1 | - | 3 | 623 | 6821724 |