Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nhsl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163592.1 | NP_001157064.1 | 7 | 4125 | ||
NM_173390.3 | NP_775566.3 | 6 | 4648 | |||
GenBank mRNA | AK043447.1 | - | 2 | 574 | ||
AK134837.1 | - | 2 | 596 | |||
AK220386.1 | - | 2 | 693 | |||
BC013565.1 | - | 2 | 247 | |||
BC061949.1 | - | 2 | 228 |