Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rccd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173445.4 | NP_775621.2 | 6 | 1680 | ||
GenBank mRNA | AK030872.1 | - | 6 | 1651 | ||
AK047578.1 | - | 825 | ||||
AK084219.1 | - | 4 | 1606 | |||
AK088482.1 | - | 5 | 1844 | |||
AK088662.1 | - | 6 | 1321 |