Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fnip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173753.4 | NP_776114.2 | 18 | 3917 | ||
GenBank mRNA | AK042779.1 | - | 5 | 1382 | ||
AK047814.1 | - | 5 | 1158 | |||
AK122574.1 | - | 18 | 3865 | |||
BC024776.1 | - | 3 | 763 | |||
BC079892.1 | - | 18 | 3907 | 30543296 |