Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gimap5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175035.5 | NP_778200.1 | 3 | 704 | ||
GenBank mRNA | AB126961.1 | - | 3 | 704 | ||
AK052694.1 | - | 3 | 637 | |||
BC095995.1 | - | 3 | 616 | 30470329 |