Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lin52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173756.4 | NP_776117.1 | 6 | 515 | ||
GenBank mRNA | AK030923.1 | - | 6 | 516 | ||
BC120496.1 | - | 6 | 601 | 40129419 | ||
BC137693.1 | - | 6 | 601 | 8860712 |