Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atg4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174874.3 | NP_777363.1 | 13 | 1677 | ||
GenBank mRNA | AK028712.1 | - | 13 | 1688 | ||
AK031570.1 | - | 13 | 1678 | |||
AK144239.1 | - | 13 | 1682 |