Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lpar6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175116.4 | NP_780325.1 | 1 | 1485 | ||
GenBank mRNA | AK008952.1 | - | 1 | 1360 | ||
AK011967.1 | - | 1 | 1375 | |||
AK032901.1 | - | 1 | 1482 | |||
BC047931.1 | - | 1 | 1361 | 5251236 | ||
BC057175.1 | - | 1 | 1348 | |||
BC145940.1 | - | 1 | 1111 | 40131243 |