Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stbd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175096.3 | NP_780305.1 | 2 | 687 | ||
GenBank mRNA | AK050416.1 | - | 2 | 688 | ||
BC052666.1 | - | 2 | 648 | |||
BC118615.1 | - | 3 | 756 | 40107322 | ||
BC118661.1 | - | 2 | 670 | 40107321 |