Search Thermo Fisher Scientific
- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Search Thermo Fisher Scientific
Gene Symbol: Rab39b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175122.6 | NP_780331.1 | 2 | 1488 | ||
GenBank mRNA | AK039013.1 | - | 2 | 1462 | ||
AK048981.1 | - | 2 | 1462 | |||
AK160737.1 | - | 2 | 1465 | |||
BC050853.1 | - | 2 | 1482 | 6517891 | ||
BC052472.1 | - | 2 | 1413 | 6833461 |