Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: March1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166372.1 | NP_001159844.1 | 6 | 525 | ||
NM_001166375.1 | NP_001159847.1 | 2 | 244 | |||
NM_001293784.1 | NP_001280713.1 | 7 | 734 | |||
NM_175188.4 | NP_780397.2 | 6 | 681 | |||
GenBank mRNA | AK036489.1 | - | 2 | 245 | ||
AK086052.1 | - | 7 | 693 | |||
AK089040.1 | - | 2 | 244 | |||
AK137337.1 | - | 6 | 481 | |||
AK152592.1 | - | 2 | 246 | |||
AK163081.1 | - | 2 | 245 | |||
BC066008.1 | - | 6 | 681 | 5703753 |