Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291002.1 | NP_001277931.1 | 10 | 1489 | ||
NM_175154.3 | NP_780363.1 | 10 | 1595 | |||
GenBank mRNA | AK012757.1 | - | 3 | 451 | ||
AK082521.1 | - | 10 | 1486 | |||
AK152851.1 | - | 8 | 1178 | |||
AK154762.1 | - | 10 | 1444 | |||
BC079843.1 | - | 10 | 1459 | 5690056 |